Easy-Squeeze Wash Bottles, 6/Cs.
Actual product may vary
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy to fill and ready to use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorChemical Label
FIS02-897-11BlueWater
FIS02-897-3BlueDeionized Water
FIS02-897-4BlueDistilled Water
FIS02-897-10NaturalUniversal
FIS02-897-5NaturalEthanol
FIS02-897-1RedAcetone
Catalog number FIS02-897-11
Price (SGD)
127.00
Case
Color:
Blue
Chemical Label:
Water
Price (SGD)
127.00
Case
Easy to fill and ready to use
  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping
Label information includes:
  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Specifications
Capacity17 oz., 500mL
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorBlue
Diameter (English)2.99 in.
IncludesPTFE membrane
MaterialLDPE
Closure ColorBlue
Certifications/ComplianceISO 13485:2003
Closure IncludedYes
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)173 mm
Product TypeWash Bottle
Height (English)6.81 in.
Chemical LabelWater
Closure MaterialPolypropylene
Closure Size38-415 mm
Diameter (Metric)76 mm
Closure TypePolypropylene Closure
Through the Closure DispensingNo