Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Actual product may vary
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Higher temperature resistance than LDPE or HDPE
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FIS02-896B60 mL28-415
Catalog number FIS02-896B
Price (SGD)
-
Capacity (Metric):
60 mL
Closure Size:
28-415
Higher temperature resistance than LDPE or HDPE
  • Handle up to 120°C (248°F)† for short periods and 100°C (212°F) continuously
  • Brittleness temperature of 0°C (32°F)
  • Rigid walls
  • Excellent chemical resistance
  • Can be sterilized by all normal methods, except dry heat and radiation
Completely disengage threads or remove cap before autoclaving.
Specifications
Capacity2 oz., 60mL
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorTranslucent
MaterialPPCO
SterilityNon-sterile
Thread Size (Metric)415
Neck Finish28 mm
Certifications/ComplianceISO 13485:2003; BSI:FM 653694
Closure IncludedYes
MouthWide
Product TypeWide Mouth Bottle
Closure LinerUnlined
Closure MaterialPPCO
Closure Size28-415
Closure TypeScrew Cap