Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Actual product may vary
Thermo Scientific
Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FIS02-893-5C125 mL12/Pk.
FIS02-893-5A30 mL12/Pk.
FKS1035350130 mL12/Pk.
2104-000260 mL72/Pk.
FIS02-893-5B60 mL72/Pk.
2104-0004125 mL12/Pk.
FKS10569831125 mL12/Pk.
2104-0008250 mL12/Pk.
FIS02-893-5D250 mL12/Pk.
FKS10623141250 mL12/Pk.
2104-0016500 mL12/Pk.
FIS02-893-5E500 mL12/Pk.
FKS10633141500 mL12/Pk.
2104-00321 L6/Pk.
FIS02-893-5F1 L6/Pk.
FKS105473411 L6/Pk.
FIS10-346-91500 mL24/Pk.
Catalog number FIS02-893-5C
Price (SGD)
112.00
Pack
Capacity (Metric):
125 mL
Quantity:
12/Pk.
Price (SGD)
112.00
Pack
Utilize Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles for countless field applications. These containers exhibit excellent chemical resistance to most acids, bases and alcohols and are suitable for freezer use to -100°C and are Leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance to most corrosives make these bottles versatile and indispensable in your lab
  • Ideal for storing, shipping, and packaging liquids — uniform walls provide durability and resistance to splitting or punctures
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
Polypropylene screw closure
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity4 oz., 125 mL
Capacity (Metric)125 mL
ColorTranslucent
DescriptionLab Quality Wide-Mouth Bottle
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Storage and Shipping of Small Volume, Light Sensitive, Valuable Reagents
MaterialHDPE
No. per Case72/Cs.
No. per Pack12/Pk.
Quantity12/Pk.
SterilityNon-sterile
Closure ColorNatural
Neck Finish38 mm
Green FeaturesACT Ecolabel (Platinum)
Closure IncludedYes
MouthWide
ACT LabelYes
Height (Metric)99 mm
Height (English)11.1 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38 mm
Diameter (Metric)50 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Capacity (English)4 oz.
ShapeRound
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
DisposableReusable
Product TypeWide Mouth Lab Quality Bottle
Unit SizePack