Kimble™ KIMAX™-35 300mL BOD Bottles
Kimble™ KIMAX™-35 300mL BOD Bottles
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™-35 300mL BOD Bottles
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Have Questions?
Catalog NumberNo. SequenceStopper
FIS02-885AUnnumberedWith stopper
Catalog number FIS02-885A
Price (SGD)
-
No. Sequence:
Unnumbered
Stopper:
With stopper
For sampling and incubation of aqueous samples for biochemical oxygen demand (BOD) analysis
Flared mouth forms a water seal to prevent air from being drawn into the bottle during incubation, while shoulder radius provides an interior shape that sweeps entrapped air out of the stopper opening.
  • Type 1, Class B borosilicate glass
  • Available numbered and unnumbered With glass stopper unless otherwise noted
  • Optional snap lid secures stopper in bottle and reduces evaporation of water seal during incubation (sold separately, 02-939)
Specifications
Certifications/ComplianceASTM E438 type I, class A
Volume (Metric)300 mL
For Use With (Application)For sampling and incubation of aqueous samples
MaterialBorosilicate Glass
No. SequenceUnnumbered
StopperWith stopper
Volume (English)10.14 oz.
Height (English)6.5 in.
Height (Metric)16.5 cm
Diameter (English)2.7 in.
Diameter (Metric)6.9 cm
Unit SizeCS