Fisherbrand™ Large-Orifice Pipet Tips
Actual product may vary
Fisherbrand™ Large-Orifice Pipet Tips
Fisherbrand™ Large-Orifice Pipet Tips helps you pipet difficult liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatVolume (Metric)ColorLength (English)
FIS02-707-134Hinged Rack1 to 200 μLYellow2 in.
FIS02-707-145Rack100 to 1000 μLNatural3.4 in.
Catalog number FIS02-707-134
Price (SGD)
-
Format:
Hinged Rack
Volume (Metric):
1 to 200 μL
Color:
Yellow
Length (English):
2 in.
Genomic/Wide Orifice assists in pipetting small volumes of viscous materials.
  • Assist in pipetting small volumes of viscous materials
  • Help eliminate shearing of genomic DNA and fragile cells
Specifications
Volume (Metric)1 to 200 μL
SterilitySterile
ColorYellow
Length (English)2 in.
Length (Metric)5cm
FormatHinged Rack
Tip StyleGenomic/Wide Orifice
PackagingTips come in a rack.
For Use With (Equipment)Costar Onepette, Eppendorf, Eppendorf Reference 2000, Eppendorf Research 2100, Eppendorf Research 3130, Eppendorf Reference 2000, Eppendorf Research 2100, Eppendorf, Thermo Scientific Finnpipette Digital
Unit SizePack