Standardization Pipet Tips
Actual product may vary
Fisherbrand
Standardization Pipet Tips
Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityColorPackaging
FIS02-681-134Non-sterileNaturalBulk
FIS02-681-140Non-sterileYellowBulk Pack
Catalog number FIS02-681-134
Price (SGD)
55.00
Pack
Sterility:
Non-sterile
Color:
Natural
Packaging:
Bulk
Price (SGD)
55.00
Pack
Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
  • Soft pliable tip collar allows this tip to fit on pipets made by various manufacturers
  • Lower cost and increased convenience by purchasing fewer products
  • Available in a variety of packaging options
  • Eliminate the need to buy pipetter-specific tips
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA and pyrogen free
Specifications
Volume (Metric)0.1 to 200 μL
SterilityNon-sterile
ColorNatural
AutoclavableAutoclavable
Length (English)2 in.
Length (Metric)5cm
Clean ClaimsDNase, RNase, DNA, PCR inhibitor, and Pyrogen Free
FormatBulk
Tip StyleUniversal
PackagingBulk
For Use With (Equipment)Universal Fit