Cover Glasses/Slips
Actual product may vary
Hausser Scientific
Cover Glasses/Slips
Crafted from high-quality borosilicate glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)For Use With (Equipment)
FIS02-671-570.5 mm25 mm60 mmSedgewick-Rafter Cell Counter
FIS02-617-5810.5 mm20 mm26 mm31xx /35xx Hemacytometers
Catalog number FIS02-671-57
Price (SGD)
109.00
Each
Thickness (Metric):
0.5 mm
Length (Metric):
25 mm
Width (Metric):
60 mm
For Use With (Equipment):
Sedgewick-Rafter Cell Counter
Price (SGD)
109.00
Each
Crafted from high-quality borosilicate glass
Specifications
Product TypeCoverslip
ShapeRectangle
MaterialBorosilicate Glass
Length (Metric)25 mm
Width (Metric)60 mm
Thickness (Metric)0.5 mm
For Use With (Equipment)Sedgewick-Rafter Cell Counter
Quantity1 Ea.
Unit SizeEach