Phase Contrast Hemacytometer
Actual product may vary
Hausser Scientific
Phase Contrast Hemacytometer
For making platelet counts
Have Questions?
Catalog NumberQuantity
FIS02-671-541 Ea.
Catalog number FIS02-671-54
Price (SGD)
715.00
Each
Quantity:
1 Ea.
Price (SGD)
715.00
Each
For making platelet counts
  • Constructed of Borosilicate Glass with Bright Line ruling
  • Counting chamber is reduced in overall thickness
  • For use with a phase contrast microscope
  • Includes two cover glasses
  • Dimensions: 32 × 75mm
Specifications
Product TypePhase Contrast Hemacytometer
Grid TypeBright Line
Capacity (Metric)0.1μL
MaterialBorosilicate Glass
Quantity1 Ea.
Depth (English)0.003 in.
Depth (Metric)0.1mm
For Use With (Application)Determining Platelet Counts, Phase Contrast Microscopy
For Use With (Equipment)Phase microscope
IncludesTwo cover glasses
Unit SizeEach