Bright Line™ Counting Chamber
Actual product may vary
Bright Line™ Counting Chamber
Bright Line™ Counting Chamber
Bright Line™ Counting Chamber
Bright Line™ Counting Chamber
Counting Chamber; Bright Line; Hausser Scientific; Molded Borosilicate Glass; Double improved Neubauer ruling
Have Questions?
Catalog number FIS02-671-51A
Price (SGD)
784.00
Each
Price (SGD)
784.00
Each
Counting Chamber; Bright Line; Hausser Scientific; Molded Borosilicate Glass; Double improved Neubauer ruling
Specifications
Product TypeHemacytometer
Grid TypeBright Line
Capacity (Metric)0.1μL
MaterialBorosilicate Glass
QuantityEach
Depth (English)0.003 in.
Depth (Metric)0.01cm
For Use With (Application)Accurate Counting of Blood Cells, Platelets, Dust, Yeast Clusters, and Other Specimens in Spinal and Other Fluids
Unit SizeEach