Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Blood, Hematology and Coagulation Testing Products
▸
Cell Counting Devices
▸
Cell Counters
Actual product may vary
Bal Supply
Cell Counters
For automatic calculation including percentages.
Have Questions?
Change view
Catalog Number
Style
FIS02-670-14
8 Counting units; 2 reset knob
FIS02-670-11
1 Counting unit; 1 reset knob
FIS02-670-12
2 Counting units; 1 reset knob
FIS02-670-13
5 Counting units; 2 reset knob
4 Options
Catalog number
FIS02-670-14
Price (SGD)
284.00
Each
Add to cart
Style:
8 Counting units; 2 reset knob
Price (SGD)
284.00
Each
Add to cart
For automatic calculation including percentages
Totaling unit rings a bell at 100 increments
Dual reset knobs
Last unit is the totaling unit, which records total number of strokes made on other keys
Removable tabs on top of counter provide area for identification of classification being counted
Specifications
Product Type
Differential Cell Counter
Displayed Text
0 to 999
Style
8 Counting units; 2 reset knob
For Use With (Application)
Cell Counting with Automatic Calculations