Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper
Actual product may vary
BD
Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper
Use for serum determinations in chemistry, serology and immunohematology.
Have Questions?
Catalog NumberDiameter (Metric) OuterCapacity (Metric)Product Type
FIS02-657-2713 x 75 mm3 mLBlood Collection Tube
Catalog number FIS02-657-27
Price (SGD)
102.00
Pack
Diameter (Metric) Outer:
13 x 75 mm
Capacity (Metric):
3 mL
Product Type:
Blood Collection Tube
Price (SGD)
102.00
Pack
  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
  • With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Specifications
Product TypeBlood Collection Tube
AdditiveClot Activator, Silicone Coated
MaterialPlastic
Closure TypeTraditional Cap
Capacity (Metric)3 mL
Closure ColorRed
Diameter (Metric) Outer13 x 75 mm
Unit SizePack