Cover Glasses/Slips
Actual product may vary
Hausser Scientific
Cover Glasses/Slips
Crafted from high-quality borosilicate glass.
Have Questions?
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)For Use With (Equipment)
FIS02-617-5810.5 mm20 mm26 mm31xx /35xx Hemacytometers
Catalog number FIS02-617-581
Price (SGD)
169.00
Each
Thickness (Metric):
0.5 mm
Length (Metric):
20 mm
Width (Metric):
26 mm
For Use With (Equipment):
31xx /35xx Hemacytometers
Price (SGD)
169.00
Each
Crafted from high-quality borosilicate glass
Specifications
Product TypeCoverslip
ShapeRectangle
MaterialBorosilicate Glass
Length (Metric)20 mm
Width (Metric)26 mm
Thickness (Metric)0.5 mm
For Use With (Equipment)31xx /35xx Hemacytometers
Quantity1 Ea.
Unit SizeEach