Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Height (Metric) | Tolerance |
|---|---|---|---|
| FIS02-617-155 | 250 mL | 23.5 cm | 0.15 mL |
| FIS02-617-152 | 25 mL | 11.5 cm | 0.04 mL |
| FIS02-617-156 | 500 mL | 27 cm | 0.25 mL |
Calibrated To Contain within Class A tolerances.
Suitable for trace element analysis, isotope separations, sample digestions, and the handling of sensitive biological solutions