Lab Pots
Lab Pots
Actual product may vary
Medegen
Lab Pots
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerCapacity (Metric)
FIS02-60333.02cm18.9 L
FIS02-60226.33cm10.8 L
Catalog number FIS02-603
Price (SGD)
560.00
EA
Add to cart
Diameter (Metric) Inner:
33.02cm
Capacity (Metric):
18.9 L
Price (SGD)
560.00
EA
Add to cart
  • Ideal containers for heating, sterilizing
  • Stainless steel to spread heat evenly
Specifications
Capacity20 qt., 18.9L
Certifications/ComplianceNSA
Diameter (English) Inner13 in.
Diameter (Metric) Inner33.02cm
For Use With (Application)Ideal containers for heating, sterilizing
Height (English)11 in.
Height (Metric)27.94cm
Temperature (English)275°F
Temperature (Metric)135°C
Capacity (English)20 qt.
Capacity (Metric)18.9 L
MaterialAluminum, Stainless Steel
Product TypeLaboratory Pot
Unit SizeEA