Lab Pots
Actual product may vary
Medegen
Lab Pots
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerCapacity (Metric)
FIS02-60226.33cm10.8 L
FIS02-60333.02cm18.9 L
Catalog number FIS02-602
Price (SGD)
407.00
Each
Diameter (Metric) Inner:
26.33cm
Capacity (Metric):
10.8 L
Price (SGD)
407.00
Each
  • Ideal containers for heating, sterilizing
  • Stainless steel to spread heat evenly
Specifications
Capacity11.5 qt., 10.8L
Certifications/ComplianceNSA
Diameter (English) Inner10.37 in.
Diameter (Metric) Inner26.33cm
For Use With (Application)Ideal containers for heating, sterilizing
Height (English)8.87 in.
Height (Metric)22.52cm
Temperature (English)275°F
Temperature (Metric)135°C
Capacity (English)11.5 qt.
Capacity (Metric)10.8 L
MaterialAluminum, Stainless Steel
Product TypeLaboratory Pot
Unit SizeEach