Nalgene™ Griffin Low-Form PFA Plastic Beakers
Actual product may vary
Nalgene™ Griffin Low-Form PFA Plastic Beakers
Nalgene™ Griffin Low-Form PFA Plastic Beakers
Nalgene™ Griffin Low-Form PFA Plastic Beakers
Thermo Scientific
Nalgene™ Griffin Low-Form PFA Plastic Beakers
Handle temperatures from -270° to +250°C using these sturdy and tough low-form PFA beakers, ideal for continual lab usage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-591-40C100 mL
FIS02-591-22A50 mL
Catalog number FIS02-591-40C
Price (SGD)
1,719.00
Each
Capacity (Metric):
100 mL
Price (SGD)
1,719.00
Each
Handle temperatures from -270° to +250°C using sturdy and tough Thermo Scientific™ Nalgene™ Griffin Low-Form PFA Beakers. Molded from chemically resistant perfluoroalkoxy resin and featuring a heavy-wall design, these beakers are ideal for continual lab usage.
  • No-drip, single-spout design for precision pouring control
  • Highly resistant to most chemicals, including the strongest sulfuric, nitric, chromic and phosphoric acids — will not contaminate contents with leachables
  • Autoclavable/PFA
  • Not for use on hotplates
Specifications
ColorNatural translucent
DescriptionBeaker Griffin Low-Form PFA 100 mL
Capacity (Metric)100 mL
MaterialPFA
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
ScaleNot Graduated
StyleGriffin, Low Form
Unit SizeEach