PYREX™ Beaker with Glass Handle
PYREX™ Beaker with Glass Handle
Actual product may vary
PYREX™ Beaker with Glass Handle
Ideal for handling hot solutions.
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-5433000 mL
Catalog number FIS02-543
Price (SGD)
1,415.00
EA
Add to cart
Capacity (Metric):
3000 mL
Price (SGD)
1,415.00
EA
Add to cart
Corning™ PYREX™ Beakers with Glass Handle are ideal for handling hot solutions.
A sturdy, molded glass handle is secured to these beakers by a stainless steel strap which fits into a recess in the beaker wall. Hot liquids can be handled easily and safely.
Specifications
Capacity (English)101.44 oz.
Diameter (English) Outer6.1 in.
Diameter (Metric) Outer15.5 cm
Capacity (Metric)3000 mL
MaterialBorosilicate Glass
For Use With (Application)Permits Safe and Burn Proof Handling of Hot Solutions
Height (English)8.38 in.
Height (Metric)21.3 cm
IncludesHandle, Spout
Quantity1/Pk.
TypeBeaker
DisposableReusable
GraduatedNo
Unit SizeEA