Kimble™ KIMAX™ GL 45 Media/Storage Bottles
Kimble™ KIMAX™ GL 45 Media/Storage Bottles
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ GL 45 Media/Storage Bottles
Perfect for mixing, storing or transporting culture media, chemicals or solvents
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS02-542D1000 mL100 to 900mL
FIS02-542A100 mL20 to 80mL
FIS02-542B250 mL50 to 200mL
FIS02-542C500 mL100 to 400mL
Catalog number FIS02-542D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
100 to 900mL
Perfect for mixing, storing or transporting culture media, chemicals or solvents
  • Autoclavable Type 1 borosilicate glass
  • Permanent white-enamel graduations and marking spots
  • Approx. 30mm I.D. GL 45 screw-thread opening
  • Linerless blue polypropylene cap autoclavable to 140°C (284°F) and replaceable, drip-free, clear polypropylene pour ring included
Specifications
Capacity33.8 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)33.8 oz.
ColorWhite
Diameter (English)3.9 in.
Diameter (English) Inner Neck1.18 in.
Diameter (Metric) Inner Neck30 mm
IncludesClear drip-free Polypropylene pouring ring
MaterialBorosilicate Glass (Type I)
Closure ColorBlue
Graduations100 to 900mL
Closure IncludedYes
AutoclavableAutoclavable
GraduatedYes
ShapeRound
Height (Metric)225 mm
Product TypeMedia Bottle
Height (English)8.8 in.
Closure MaterialPolypropylene
Closure SizeGL 45
Diameter (Metric)101 mm
Closure TypeScrew Cap
Unit SizeCS