▸
Histology and Cytology
▸
Tissue Grinders
▸
Disposable Tissue Grinders
Actual product may vary
Disposable Tissue Grinders
Closed tissue grinder system minimizes sample contact by aerosols and sample overflow
Have Questions?
Catalog Number
Capacity
Product Type
Autoclavable
FIS02-542-11
1.69 oz., 50mL
Ultra Tissue Grinder
Autoclavable
Catalog number
FIS02-542-11
Price (SGD)
-
Capacity:
1.69 oz., 50mL
Product Type:
Ultra Tissue Grinder
Autoclavable:
Autoclavable
Sterile and pre-assembled
Closed system prevents risk of contact with sample
Easy-to-open Tyvek™ packaging
Extra cap and patient label provided
Standard Tissue Grinders
Feature a molded-in abrasive surface; no sediment to obscure sample
Available in 15, 35 and 50mL sizes
Ultra Tissue Grinders
Silica cast tip provides enhanced grinding for difficult samples
Available in 15 and 50mL sizes
Specifications
Capacity (English)
1.69 oz.
For Use With (Application)
Grinding small tissue samples
Autoclavable
Autoclavable
Disposable
Single-use
Includes
Polypropylene tube, polystyrene pestle and tip, HDPE cap and extra cap
Capacity (Metric)
50 mL
Capacity
1.69 oz., 50mL
Sterility
Sterile
Product Type
Ultra Tissue Grinder