Disposable Tissue Grinders
Actual product may vary
Disposable Tissue Grinders
Disposable Tissue Grinders
Disposable Tissue Grinders
Closed tissue grinder system minimizes sample contact by aerosols and sample overflow
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityProduct TypeAutoclavable
FIS02-542-090.5 oz., 15mLTissue GrinderNot Autoclavable
FIS02-542-111.69 oz., 50mLUltra Tissue GrinderAutoclavable
FIS02-542-071.69 oz., 50mLTissue GrinderAutoclavable
Catalog number FIS02-542-09
Price (SGD)
-
Capacity:
0.5 oz., 15mL
Product Type:
Tissue Grinder
Autoclavable:
Not Autoclavable
  • Sterile and pre-assembled
  • Closed system prevents risk of contact with sample
  • Easy-to-open Tyvek™ packaging
  • Extra cap and patient label provided
Standard Tissue Grinders
  • Feature a molded-in abrasive surface; no sediment to obscure sample
  • Available in 15, 35 and 50mL sizes
Ultra Tissue Grinders
  • Silica cast tip provides enhanced grinding for difficult samples
  • Available in 15 and 50mL sizes
Specifications
Capacity (English)0.5 oz.
For Use With (Application)Grinding small tissue samples
AutoclavableNot Autoclavable
DisposableSingle-use
IncludesPolypropylene tube, vitrified pestle tip, HDPE cap and extra cap
Capacity (Metric)15 mL
Capacity0.5 oz., 15mL
SterilitySterile
Product TypeTissue Grinder
Unit SizeCase