PYREX™ Low Form Griffin Beakers
PYREX™ Low Form Griffin Beakers
Actual product may vary
Fisherbrand
PYREX™ Low Form Griffin Beakers
Offers optimum balance between thermal shock resistance and mechanical strength
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-540R2000 mL
FIS02-540C10 mL
FIS02-540E20 mL
FIS02-540G50 mL
FIS02-540H100 mL
FIS02-540J150 mL
FIS02-540K250 mL
FIS02-540L400 mL
FIS02-540M600 mL
FIS02-540P1 L
FIS02-540S3000 mL
FIS02-540T4000 mL
Catalog number FIS02-540R
Price (SGD)
356.00
PK
Add to cart
Capacity (Metric):
2000 mL
Price (SGD)
356.00
PK
Add to cart
With pouring spout and extra-large marking spot
Corning™ PYREX™ Low Form Griffin Beakers offer optimum balance between thermal shock resistance and mechanical strength.
With pouring spout and extra-large marking spot.
Beakers, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10 mL size is not graduated.
Specifications
Capacity (English)67.62 oz.
Diameter (English) Outer5.15 in.
Diameter (Metric) Outer13.1 cm
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
For Use With (Application)For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
Graduation Range200 to 1800 mL
Height (English)7.59 in.
Height (Metric)19.3 cm
IncludesSpout
Increments100 mL
Label TypeWhite Marking
Quantity4 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual White Metric Scale
StyleGriffin
Unit SizePK