Metric Economical Stainless Steel Calibration Weight Sets, Class 7
Actual product may vary
Troemner
Metric Economical Stainless Steel Calibration Weight Sets, Class 7
Economical, quality weights
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMassIncludesQuantityShape
FIS02-301-10710 g to 50 gCylindrical weight: 50g, 20g, 2 x 10g, 5g, 2 x 2g, 1g, 303 Stainless Steel sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, Plastic case16/SetCylindrical with Handling Knob
FIS02-301-10210 mg to 100 gCylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, 303 Stainless Steel sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, Plastic case17/SetCylindrical with Handling Knob
Catalog number FIS02-301-107
Price (SGD)
235.00
Each
Mass:
10 g to 50 g
Includes:
Cylindrical weight: 50g, 20g, 2 x 10g, 5g, 2 x 2g, 1g, 303 Stainless Steel sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, Plastic case
Quantity:
16/Set
Shape:
Cylindrical with Handling Knob
Price (SGD)
235.00
Each
  • Sets designed to achieve any total mass by combining smaller masses
  • Weights 1g to 100g are of one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight
  • Weights 200g through 2kg have an adjusting cavity on the top that is sealed with a plug
Specifications
Product TypeMetric Economical Weight Set
ClassificationASTM Class 7
MaterialStainless Steel
Weight (Metric)10 mg to 50 g
Density7.85g/cm3 at 20°C
Mass10 g to 50 g
ShapeCylindrical with Handling Knob
IncludesCylindrical weight: 50g, 20g, 2 x 10g, 5g, 2 x 2g, 1g, 303 Stainless Steel sheet weight: 500mg, 2 x 200mg, Aluminum sheet weight: 100mg, 50mg, 2 x 20mg, 10mg, Plastic case
Quantity16/Set
Packaging QuantityMultiple
Unit SizeEach