Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Balances and Scales
▸
Calibration Weights
▸
Weight Set Case
Actual product may vary
Troemner
Weight Set Case
Polycarbonate weight set case for Troemner ASTM weights
Have Questions?
Catalog number
FIS02-216-360
Price (SGD)
514.00
Each
Add to cart
Price (SGD)
514.00
Each
Add to cart
Case holds ASTM Weight Set: 500g-1mg
Designed for: screw knob
Material: polycarbonate
Dimensions: 8L x 7W x 3.5 in.H (20.3 x 17.8 x 8.9cm)
Protective urethane lid bumper for each weight
Internal lid to protect mg weights
Red, color-coded insert
Specifications
Product Type
Analytical Weight Set Case
Material
Polycarbonate