Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Clamps
▸
Talboys™ Heavy Duty Holder
Actual product may vary
Troemner
Talboys™ Heavy Duty Holder
Comprised of a sturdy aluminum alloy with a brushed finish. The Troemner™ Talboys™ Heavy Duty Holder is made in the USA.
Have Questions?
Catalog Number
Type
FIS02-216-354
Heavy Duty Holder
Catalog number
FIS02-216-354
Price (SGD)
445.00
Each
Add to cart
Type:
Heavy Duty Holder
Price (SGD)
445.00
Each
Add to cart
Accommodates all shafts 1/4in. (6.4mm) to 1in. (25.4mm) diameter
Oversized, easy to grip knobs for precise tightening
Supports rods on a 4in. (10.2cm) long surface to prevent vibrations
Specifications
Type
Heavy Duty Holder
Description
Accommodates all shafts 1/4 in. (6.4 mm) to 1 in. (25.4 mm) diameter