Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Stands and Rings
▸
Talboys™ Labjaws™ Open Ring Clamps
Actual product may vary
Troemner
Talboys™ Labjaws™ Open Ring Clamps
Ideal for supporting funnels, round bottom flasks, reaction vessels, and other apparatus that requires lower support
Have Questions?
Catalog Number
Length (Metric)
Length (English)
Diameter (Metric) Outer
Diameter (English) Outer
FIS02-216-326
32.8 cm
12.91 in.
72 mm
3 in.
Catalog number
FIS02-216-326
Price (SGD)
315.00
Each
Add to cart
Length (Metric):
32.8 cm
Length (English):
12.91 in.
Diameter (Metric) Outer:
72 mm
Diameter (English) Outer:
3 in.
Price (SGD)
315.00
Each
Add to cart
Opening in PVC coated aluminum ring allows for easy removal of sample container
PVC coating protects glassware
Long extension arm permits depth adjustment of the open ring from the lab-frame or ring stand
Specifications
Material
PVC
Length (English)
12.91 in.
Length (Metric)
32.8 cm
Diameter (English) Outer
3 in.
Diameter (Metric) Outer
72 mm
Product Type
Open Extension Ring