Talboys™ Labjaws™ Support Stand for Double Buret Clamp
Support stand with standard rod for use with the nickel-plated Double Buret Clamp (sold separately). Clamp unit attaches to the support rod with built-in reinforced hook connector. Support rod attaches to the porcelain base. Rod (Dia x L): 1.3 x 57.8cm, Stand (L x W x H): 17.8 x 33 x 2.5cm
Have Questions?
Catalog Number
For Use With (Equipment)
FIS02-216-298
Labjaws
Catalog number FIS02-216-298
Price (SGD)
813.00
Each
For Use With (Equipment):
Labjaws
Price (SGD)
813.00
Each
For use with the nickel-plated Double Buret Clamp (sold separately), which holds any size buret from micro to 100mL capacity