Talboys™ Ultrajaws Heavy-Duty Clamps
Actual product may vary
Troemner
Talboys™ Ultrajaws Heavy-Duty Clamps
Enables secure gripping and positioning with added strength and durability
Have Questions?
Catalog NumberSizeSize (English) GripLength (English) Arm
FIS02-216-239Large4.06 in.5 in.
Catalog number FIS02-216-239
Price (SGD)
272.00
Each
Size:
Large
Size (English) Grip:
4.06 in.
Length (English) Arm:
5 in.
Price (SGD)
272.00
Each
Enables secure gripping and positioning with added strength and durability
  • Three-pronged grip with large grip adjustment range
  • Closed yoke construction that minimizes contamination and corrosion of internal components
  • Precise pressure regulation when gripping glassware surfaces reduces the chance of breakage
  • Constructed of nickel-plated zinc with vinyl sleeves
  • For temperatures above 100°C, fiberglass covers
An additional holder must be purchased in order to attach clamps to frames or other apparatus.
Specifications
TypeHeavy Duty Clamp
MaterialNickel, Zinc
For Use With (Application)Use in holding and securing Glassware with precision ground Glass joints
Length (English) Arm5 in.
Length (Metric) Arm12.7 cm
SizeLarge
Size (Metric) Grip10.3 cm
Size (English) Grip4.06 in.
Unit SizeEach