Stainless Steel Lab Frame Rod
Actual product may vary
Stainless Steel Lab Frame Rod
Heavy-duty rods with maximum strength and durability.
Have Questions?
Catalog NumberLength (Metric)
FIS02-215-48091.4 cm
Catalog number FIS02-215-480
Price (SGD)
102.00
Each
Length (Metric):
91.4 cm
Price (SGD)
102.00
Each
  • Made of 303 stainless steel
  • Diameter: 13mm (0.51 in.)
  • Length: 91.4cm (36 in.)
Specifications
Length (English)36 in.
Length (Metric)91.4 cm
Unit SizeEach