Alloy 8 Metric Precision Weight Sets, Class 1
Actual product may vary
Troemner
Alloy 8 Metric Precision Weight Sets, Class 1
Excellent for high-precision calibrations and reference standards
Have Questions?
Catalog NumberQuantity
FIS02-215-11522/Set
Catalog number FIS02-215-115
Price (SGD)
4,633.00
Each
Quantity:
22/Set
Price (SGD)
4,633.00
Each
Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2 (2mg, 20mg, etc.) and one weight for all other inclusive values
  • Certificate of calibration is available; contact your Fisher Customer Service Representative
Specifications
Product TypeMedium Metric Analytical Weight Set
ClassificationASTM Class 1
MaterialStainless Steel
Weight (Metric)1 mg to 200 g
Mass200 g, 100 g, 50 g, (2)20 g, 10 g, 5 g, (2)2 g, 1 g, 500 mg, (2)200 mg, 100 mg, 50 mg, (2)20 mg, 10 mg, 5 mg, (2)2 mg, 1 mg
IncludesStainless Steel cylindrical weight: 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminum sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Quantity22/Set
Packaging QuantityMultiple
Unit SizeEach