Hexagonal Polystyrene Weighing Dishes
Actual product may vary
Hexagonal Polystyrene Weighing Dishes
Hexagonal Polystyrene Weighing Dishes are used for handling solids or liquids during weighing. Design provides greater balance, protection and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (Metric)Diameter (Metric) Inner BaseDiameter (Metric) Inner TopWeight (Metric)
FIS02-202-10332 mm92 mm132 mm7.1 g
FIS02-202-10115 mm47 mm67 mm1.4 g
FIS02-202-10222 mm85 mm110 mm4 g
FIS02-202-1009.5 mm24 mm36 mm0.6 g
Catalog number FIS02-202-103
Price (SGD)
79.00
Pack
Depth (Metric):
32 mm
Diameter (Metric) Inner Base:
92 mm
Diameter (Metric) Inner Top:
132 mm
Weight (Metric):
7.1 g
Price (SGD)
79.00
Pack
Fisherbrand™ Hexagonal Polystyrene Weighing Dishes are used for handling solids or liquids during weighing. Design provides greater balance, protection and safety.
  • Bend easily into pouring spouts to enable nonspill transfer
  • Flat bottoms and sloping sides with 0.187 in. (4.8mm) inside corner radius ensure complete transfer
  • Do not react to most substances
  • Disposable Stack compactly for convenient storage and accessibility
Specifications
Bottom ShapeFlat
DisposableSingle-use
MaterialPolystyrene
ShapeHexagonal
TypeWeighing Dish
For Use With (Application)Design provides greater balance, protection and safety
Depth (Metric)32 mm
Diameter (Metric) Inner Base92 mm
Diameter (Metric) Inner Top132 mm
Diameter (Metric)132 (Inner Top) mm, 92 mm (Inner Base)
Weight (Metric)7.1 g
Unit SizePack