Analytical Scoop and Counterweight
Analytical Scoop and Counterweight
Actual product may vary
Analytical Scoop and Counterweight
Accessory for many mechanical and digital balances
Have Questions?
Catalog NumberIncludesLength (English)Width (English)
FIS02-190A4g. counterweight, scoop, pouring spout and storage box2 in.1.25 in.
Catalog number FIS02-190A
Price (SGD)
-
Includes:
4g. counterweight, scoop, pouring spout and storage box
Length (English):
2 in.
Width (English):
1.25 in.
Includes scoop with pouring spout, counterweight, and storage box.
Specifications
Product TypeAnalytical Scoop
MaterialAluminum
Length (English)2 in.
Width (English)1.25 in.
For Use With (Application)Quick, easy, and accurate weighing with balances
For Use With (Equipment)Mechanical and digital balances
Includes4g. counterweight, scoop, pouring spout and storage box
QuantityEach
Unit SizeEA