Moisture Analyzer Sample Pans
Actual product may vary
Mettler Toledo
Moisture Analyzer Sample Pans
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Have Questions?
Catalog NumberMaterialFor Use With (Equipment)
FIS01-913-97BStainless SteelLJ16 Infrared Moisture Analyzer
Catalog number FIS01-913-97B
Price (SGD)
263.00
Each
Material:
Stainless Steel
For Use With (Equipment):
LJ16 Infrared Moisture Analyzer
Price (SGD)
263.00
Each
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
  • 3.75 in. (96mm) in diameter
Specifications
AutoclavableAutoclavable
Bottom ShapeFlat
ColorSilver
DisposableSingle-use
MaterialStainless Steel
ShapeRound
SterilityNon-sterile
TypeMoisture Pan
For Use With (Equipment)LJ16 Infrared Moisture Analyzer
Height (Metric)15 mm
Diameter (Metric)88.9 mm
Weight (Metric)50 g
Unit SizeEach