▸
pH and Electrochemistry
▸
pH and Electrochemistry Meter Accessories
▸
Benchtop Titrator Accessories
▸
Polypropylene Titration Cups for Mettler Toledo Titrators
Actual product may vary
Mettler Toledo
Polypropylene Titration Cups for Mettler Toledo Titrators
Accessory for use with Mettler Toledo Titrators.
Have Questions?
Catalog Number
For Use With (Equipment)
FIS01-912-342
Titrator
Catalog number
FIS01-912-342
Price (SGD)
1,375.00
Case
Add to cart
For Use With (Equipment):
Titrator
Price (SGD)
1,375.00
Case
Add to cart
For use with Mettler Toledo Titrators.
Specifications
Type
Titration Cup
For Use With (Application)
Titration
Material
Plastic
Volume (Metric)
100 mL
For Use With (Equipment)
Titrator