Analytical Individual Weights: ASTM™ Class 1
Actual product may vary
Mettler Toledo
Analytical Individual Weights: ASTM™ Class 1
Low magnetic permeability and high corrosion resistance for reliable stable values
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMassProduct TypeIncludes
FIS01-912-1882 kgTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-1481 mgAnalytical Sheet WeightPlastic box; Without certificate
FIS01-912-1691 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-1702 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-1505 mgAnalytical Sheet WeightPlastic box; Without certificate
FIS01-912-1715 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-15110 mgAnalytical Sheet WeightPlastic box; Without certificate
FIS01-912-17210 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-17320 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-17450 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-154100 mgAnalytical Sheet WeightPlastic box; Without certificate
FIS01-912-175100 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-176200 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-177500 mgAnalytical Sheet WeightPlastic box; With certificate
FIS01-912-1571 gTwo-Piece Analytical WeightPlastic box; Without certificate
FIS01-912-1781 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-1792 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-1805 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-18110 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-16120 gTwo-Piece Analytical WeightPlastic box; Without certificate
FIS01-912-18220 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-16250 gTwo-Piece Analytical WeightPlastic box; Without certificate
FIS01-912-18350 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-163100 gTwo-Piece Analytical WeightPlastic box, No certificate
FIS01-912-184100 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-164200 gTwo-Piece Analytical WeightPlastic box; Without certificate
FIS01-912-185200 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-165500 gTwo-Piece Analytical WeightPlastic box; Without certificate
FIS01-912-186500 gTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-1871 kgTwo-Piece Analytical WeightPlastic box; With certificate
FIS01-912-1895 kgTwo-Piece Analytical WeightPlastic box; With certificate
Catalog number FIS01-912-188
Price (SGD)
4,663.00
Each
Mass:
2 kg
Product Type:
Two-Piece Analytical Weight
Includes:
Plastic box; With certificate
Price (SGD)
4,663.00
Each
Low magnetic permeability and high corrosion resistance for reliable stable values
  • ASTM Class 1
  • Premium stainless-steel, corrosion-resistant
  • Two-piece screw-knob construction
Specifications
Product TypeTwo-Piece Analytical Weight
Certifications/ComplianceWith certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard, A2LA accredited lab Worthington, OH
ClassificationASTM Class 1
MaterialStainless Steel
Weight (Metric)2 kg
Density7.9kg/dm3 at 20°C
Mass2 kg
ShapeCylindrical with Handling Knob
IncludesPlastic box; With certificate
QuantityEach
Packaging Quantity1
Unit SizeEach