Autoclavable Waste Bags: Plain
Autoclavable Waste Bags: Plain
Actual product may vary
Autoclavable Waste Bags: Plain
Plain style (without print).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Capacity (English)
FIS01-830ERed48 x 37 in. (122 x 94 cm)44 gal.
FIS01-826-1Clear12 x 8 in. (30 x 20 cm)1 to 2 gal.
FIS01-826-3Clear12 x 10 in. (30 x 25 cm)1 to 2 gal.
FIS01-826-4Clear10 x 15 in. (25 x 38 cm)1 to 2 gal.
FIS01-826-7Clear36 x 24 in. (91 x 61 cm)13 to 16 gal.
FIS01-830ARed12 x 8 in. (30 x 20 cm)1 to 2 gal.
FIS01-830BRed19 x 14 in. (48 x 36 cm)3 to 4 gal.
FIS01-830DRed35 x 25 in. (89 x 64 cm)13 to 16 gal.
Catalog number FIS01-830E
Price (SGD)
735.00
PK
Add to cart
Color:
Red
Dimensions (L x W):
48 x 37 in. (122 x 94 cm)
Capacity (English):
44 gal.
Price (SGD)
735.00
PK
Add to cart
Plain style bags
  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures
Specifications
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
Capacity (English)44 gal.
ColorRed
Dimensions (L x W)48 x 37 in. (122 x 94 cm)
Length (Metric)122 cm
Width (Metric)94 cm
AutoclavableAutoclavable
Width (English)37 in.
Length (English)48 in.
Thickness2 mil
MaterialPolypropylene
Temperature (Metric)140°C
Unit SizePK