Autoclavable Waste Bags: Printed
Autoclavable Waste Bags: Printed
Actual product may vary
Fisherbrand
Autoclavable Waste Bags: Printed
Printed style.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Capacity (English)Color
FIS01-828E48 x 37 in. (122 x 94 cm)44 gal.Red
FIS01-826A24 x 12 in. (61 x 30 cm)2 to 3 gal.Clear
FIS01-826C36 x 24 in. (91 x 61 cm)13 to 16 gal.Clear
FIS01-826D38 x 31 in. (97 x 79 cm)20 to 30 gal.Clear
FIS01-826E48 x 37 in. (122 x 94 cm)44 gal.Clear
FIS01-826B30 x 24 in. (76 x 61 cm)12 to 14 gal.Clear
FIS01-828A12 x 8 in. (30 x 20 cm)1 to 2 gal.Red
FIS01-828B19 x 14 in. (48 x 36 cm)3 to 4 gal.Red
FIS01-828C23 x 19 in. (58 x 48 cm)7 to 10 gal.Red
FIS01-828D35 x 25 in. (89 x 64 cm)13 to 16 gal.Red
Catalog number FIS01-828E
Price (SGD)
555.00
PK
Add to cart
Dimensions (L x W):
48 x 37 in. (122 x 94 cm)
Capacity (English):
44 gal.
Color:
Red
Price (SGD)
555.00
PK
Add to cart
Printed style bags
  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures
Specifications
AutoclavableAutoclavable
Capacity (English)44 gal.
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
ColorRed
IncludesPrinted in English and Spanish
MaterialPolypropylene
Temperature Range (Metric)140°C
Thickness (English)2 mil
Dimensions (L x W)48 x 37 in. (122 x 94 cm)
Length (English)48 in.
Length (Metric)122 cm
Width (English)37 in.
Width (Metric)94 cm
Unit SizePK