Autoclavable HD Biohazard Waste Bags
Actual product may vary
Fisherbrand
Autoclavable HD Biohazard Waste Bags
Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Thickness
FIS01-827CClear with Red Print30 x 24 in. (76 x 61 cm)1.50 mil
FIS01-814-1Natural24 x 12 in. (61 x 30 cm)1.50 mil
FIS01-814-3Natural24 x 36 in. (61 x 91 cm)1.50 mil
FIS01-827AClear with Red Print12 x 8 in. (30 x 20 cm)1.50 mil
FIS01-827DClear with Red Print36 x 24 in. (91 x 61 cm)1.50 mil
FIS01-827BClear with Red Print24 x 12 in. (61 x 30 cm)1.50 mil
FIS01-829ARed with Black Print12 x 8 in. (30 x 20 cm)1.50 mil
FIS01-829BRed with Black Print19 x 14 in. (48 x 36 cm)1.50 mil
FIS01-829CRed with Black Print23 x 19 in. (58 x 48 cm)1.50 mil
FIS01829CRed with Black Print23 x 19 in. (58 x 48 cm)1.50 mil
FIS01-829DRed with Black Print25 x 35 in. (64 x 89 cm)1.50 mil
FIS01-829FRed with Black Print30 x 33 in. (76 x 84 cm)1.50 mil
FIS01-831BRed with Black Print48 x 38 in. (122 x 97 cm)4 mil
FIS01-829ERed with Black Print48 x 38 in. (122 x 97 cm)1.50 mil
Catalog number FIS01-827C
Price (SGD)
181.00
Pack
Color:
Clear with Red Print
Dimensions (L x W):
30 x 24 in. (76 x 61 cm)
Thickness:
1.50 mil
Price (SGD)
181.00
Pack
Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.
  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
ColorClear with Red Print
Dimensions (L x W)30 x 24 in. (76 x 61 cm)
Length (Metric)76 cm
Width (Metric)61 cm
AutoclavableAutoclavable
IncludesPrinted in English and Spanish
Width (English)24 in.
Length (English)30 in.
Thickness1.50 mil
MaterialPolyethylene
Product TypeAutoclavable Bags
Temperature (Metric)127°C
Unit SizePack