Zipper Seal Sample Bags
Zipper Seal Sample Bags
Actual product may vary
Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Thickness (English)Material
FIS01-816F26 x 9 in. (66 x 23cm)4 milPolyethylene
FIS01-816-1A3 x 2 in. (7 x 5cm)2 milLDPE
FIS01-816-1B5 x 3 in. (13 x 7cm)2 milLDPE
FIS01-816C18 x 9 in. (46 x 23cm)4 milPolyethylene
FIS01-816-1C6 x 4 in. (15 x 10cm)2 milPolyethylene
FIS01-816-1D8 x 5 in. (20 x 13cm)2 milPolyethylene
FIS01-816A6 x 4 in. (15.2 x 10.16cm)4 milLDPE
FIS01-816E13 x 9 in., (33 x 23cm)4 milLDPE
FIS01-816-1E12 x 9 in. (30 x 23cm)2 milPolyethylene
FIS01-816D8 x 5 in. (20 x 13cm)4 milLDPE
Catalog number FIS01-816F
Price (SGD)
152.00
PK
Add to cart
Dimensions (L x W):
26 x 9 in. (66 x 23cm)
Thickness (English):
4 mil
Material:
Polyethylene
Price (SGD)
152.00
PK
Add to cart
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
AutoclavableNot Autoclavable
Capacity (English)5 lb.
Closure TypeZip Seal
ColorClear
MaterialPolyethylene
No. per Case200
No. per Pack50
Thickness (English)4 mil
Dimensions (L x W)26 x 9 in. (66 x 23cm)
Length (English)26 in.
Length (Metric)66 cm
Width (English)9 in.
Width (Metric)23 cm
Unit SizePK