Clavies™ High-Temperature Autoclave Bags
Clavies™ High-Temperature Autoclave Bags
Actual product may vary
Bel-Art
Clavies™ High-Temperature Autoclave Bags
Withstand high temperatures to reduce autoclave time.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
FIS01-815-3B15 x 10 in. (38.1 x 25.4cm)
FIS01-815-3A10 x 10 in.
FIS01-815-724 x 12 in. (61 x 30 cm)
Catalog number FIS01-815-3B
Price (SGD)
270.00
PK
Add to cart
Dimensions (L x W):
15 x 10 in. (38.1 x 25.4cm)
Price (SGD)
270.00
PK
Add to cart
Withstand high temperatures to reduce autoclave time
  • Suitable for discarding Petri dishes, membrane filters, multi-well culture plates, pipets and more
  • Rugged, transparent, 0.05 mm (2 mil) thick polypropylene
  • Steam autoclavable at 275°F (135°C)
Specifications
AutoclavableAutoclavable
Capacity (English)1.5 gal.
Certifications/ComplianceDart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Closure TypeNone
ColorClear
DisposableSingle-use
MaterialPolypropylene
SterilityNon-sterile
Thickness (Metric)0.050 mm
Thickness (English)2 mil
Dimensions (L x W)15 x 10 in. (38.1 x 25.4cm)
Length (English)15 in.
Length (Metric)38.1 cm
Width (English)10 in.
Width (Metric)25.4 cm
Product TypeAutoclave Bags
Unit SizePK