Write-On Scoop Sterile Sample Bag
Actual product may vary
Nasco
Write-On Scoop Sterile Sample Bag
Sterile scoop already contained inside the bag eliminates the need to carry two items to collect a sample and minimizes contamination.
Have Questions?
Catalog NumberCapacity (Metric)
FIS01-812-78523 mL
Catalog number FIS01-812-78
Price (SGD)
442.00
Pack
Capacity (Metric):
523 mL
Price (SGD)
442.00
Pack
Sterile scoop is already contained inside the bag, eliminating the need to carry two items to collect the sample and minimizing contamination. The 2 oz. (60 cc) polypropylene scoop measures 7 in. (18.5 cm) long.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate, bags should not be used at temperatures above 180°F (82°C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346°F (-210°C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Specifications
AutoclavableNot Autoclavable
Closure TypeTape and Wire
ColorClear with White Block
DisposableSingle-use
MaterialPolyethylene
SterilitySterile
Fill Volume18 oz.
Latex FreeYes
No. per Pack50/Pk.
Outer PackagingCardboard
Protective Inner PackagingPlastic
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)2.50 mil
Dimensions (L x W)23 x 11.5 cm (9 x 4.5 in.)
Length (English)9 in.
Width (English)4.5 in.
Product TypeSample Bag
Capacity (English)18 oz.
Capacity (Metric)523 mL
Length (Metric)23 cm
Width (Metric)11.5 cm
Thickness (Metric)0.064 mm
Unit SizePack