Write-On Thio Sample Bag
Actual product may vary
Nasco
Write-On Thio Sample Bag
Thio bag with write-on strip contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
Have Questions?
Catalog NumberThickness (English)Length (English)Capacity (English)
FIS01-812-5F2.25 mil7 in.4 oz.
Catalog number FIS01-812-5F
Price (SGD)
126.00
Pack
Thickness (English):
2.25 mil
Length (English):
7 in.
Capacity (English):
4 oz.
Price (SGD)
126.00
Pack
Thio bag with write-on strip contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
Unbreakable, sterile bags to collect and dechlorinate water and wastewater samples for bacteriological examination in compliance with EPA and APHA requirements.
  • Economical and easy to dispose
  • 4 oz. (118 mL) bag contains one nontoxic, non-nutritive pill impregnated with 10 mg sodium thiosulfate and has 100 mL fill-line
  • 18 oz. (532 mL) bag contains one 25 mg sodium thiosulfate pill and has 300 mL fill-line
Specifications
DescriptionStandard Whirl-Pak Thio-Bags
AutoclavableNot Autoclavable
Closure TypeTape and Wire
ColorClear
DisposableSingle-use
For Use With (Application)Sampling Chlorinated Water
IncludesContains one 25 mg sodium thiosulfate pill.
MaterialPolyethylene
SterilitySterile
Fill Volume4 oz.
Latex FreeYes
No. per Pack100/Pk.
Outer PackagingCardboard
Protective Inner PackagingPlastic
Quantity100/Pk.
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)2.25 mil
Dimensions (L x W)18 x 7.6 cm (7 x 3 in.)
Length (English)7 in.
Width (English)3 in.
Product TypeLiquid Sample Bag
Capacity (English)4 oz.
Capacity (Metric)118 mL
Length (Metric)18 cm
Width (Metric)7.6 cm
Thickness (Metric)0.057 mm
Unit SizePack