Write-On Homogenizer Sterilized Filter Bag
Actual product may vary
Nasco
Write-On Homogenizer Sterilized Filter Bag
Full-page filters solids out of a sample when using a homogenizer blender. Bags contain a finely perforated middle layer to filter solids and feature puncture proof tabs that help form a leak free closure.
Have Questions?
Catalog NumberCapacity (Metric)Closure Type
FIS01-812-51627 mLTape and Wire
Catalog number FIS01-812-5
Price (SGD)
857.00
Pack
Capacity (Metric):
1627 mL
Closure Type:
Tape and Wire
Price (SGD)
857.00
Pack
Full-page filters solids out of a sample when using a homogenizer blender. Bags contain a finely perforated middle layer to filter solids and feature puncture proof tabs that help form a leak free closure.
  • Ideal for liquid, semi-solid, or solid samples
  • Filter layer separates liquid and solids, allowing for easy pipetting
  • Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times
  • White write-on area
  • Suitable for use with most homogenizer blenders
  • Puncture proof tabs with no sharp points to damage bags or gloves
  • Collect, blend and incubate samples all in one container, lower CO2 impact to sampling bottle
  • Polyethylene
Specifications
DescriptionWith closure
AutoclavableNot Autoclavable
Certifications/ComplianceFDA Approved
Closure TypeTape and Wire
ColorClear with White Block
DisposableSingle-use
For Use With (Application)Collect, Store, and Transport Samples
For Use With (Equipment)Stomacher, Blender, Homogenizer
IncludesBag
MaterialPolyethylene
SterilitySterile
Tubing StyleContinuous perimeter seal with tape and wire closure, full-page filter
Fill Volume55 oz.
Latex FreeYes
No. per Pack250/Pk.
Outer PackagingCardboard
Product LineBlender Bag
Protective Inner PackagingPlastic
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)4 mil
Dimensions (L x W)12 x 7.5 in. (30 x 19 cm)
Length (English)12 in.
Width (English)7.5 in.
Product TypeBlender Bag (Straight Bottom)
Capacity (English)55 oz.
Capacity (Metric)1627 mL
Length (Metric)30 cm
Width (Metric)19 cm
Thickness (Metric)0.102 mm
Unit SizePack