Check Order Status
Kimble™ KIMAX™ Specific Gravity Bottle for Oils, Tars
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Specific Gravity Bottle for Oils, Tars
Determines specific gravity of various oils, tars, cements, varnishes
Have Questions?
Catalog Number
Capacity (Metric)
FIS01-710
25 mL
Catalog number
FIS01-710
Price (SGD)
839.00
PK
Add to cart
Capacity (Metric):
25 mL
Price (SGD)
839.00
PK
Add to cart
Hubbard-style bottle with standard taper 24/12 ground-glass stopper Has 0.06 in. (1.6mm) opening
Specifications
Capacity (Metric)
25 mL
Diameter (English)
1.06 in.
Material
Borosilicate Glass
Standard Taper Joint
24/12
Weight (Metric)
35g
Weight (English)
0.07 lb.
Height (Metric)
82 mm
Diameter (Metric)
27 mm