Tyvek™ 400 Hooded Coverall
Actual product may vary
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
Tyvek™ 400 Hooded Coverall
DuPont
Tyvek™ 400 Hooded Coverall
Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash.
Have Questions?
Catalog NumberSize
FIS01-361-7CLarge
Catalog number FIS01-361-7C
Price (SGD)
712.00
Case
Size:
Large
Price (SGD)
712.00
Case
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialTyvek
ColorWhite
DisposableLimited-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeRespirator Fit
Closure TypeFront Zipper
Seam TypeSerged
SterilityNon-sterile
ResistanceParticle, Non-toxic Particulates
Certifications/ComplianceUSDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Latex FreeYes
ModelTY127SWH
Size (English) Chest39.75 to 43.25 in.
Size (English) Inseam29.5 in.
Size (Metric) Chest100.96 to 109.85 cm
Size (Metric) Inseam74.93 cm
Thickness5.7 mil
For Use With (Application)Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Length (English) Sleeve35.5 in.
Length (Metric) Sleeve90.17 cm
Height (English)5.5 to 5.9 ft.
Height (Metric)1.67 to 1.8 m
Quantity25/Cs.
Unit SizeCase