KleenGuard™ A20 Breathable Particle Protection Lab Coats
Actual product may vary
Ansell
KleenGuard™ A20 Breathable Particle Protection Lab Coats
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
Have Questions?
Catalog NumberSizeClosure TypeNo. of PocketsCuff Style
FIS01-349CLargeSnaps2Open
Catalog number FIS01-349C
Price (SGD)
497.00
Case
Size:
Large
Closure Type:
Snaps
No. of Pockets:
2
Cuff Style:
Open
Price (SGD)
497.00
Case
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
  • Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
  • Knee-length, serged seams
  • Long sleeves, notched-lapel collar
  • Air and moisture vapor can pass through to aid wearer comfort
  • Meets NFPA 99 criteria for antistatic materials
  • Two styles to choose from:
    • Open wrists with pockets, snap closure
    • Elastic wrists without pockets, hook and loop closure
Specifications
Product TypeGeneral Purpose Lab Coat
SizeLarge
GenderUnisex
MaterialSMS
ColorWhite
Closure TypeSnaps
Cuff StyleOpen
DisposableLimited-use
SterilityNon-sterile
ResistanceParticulates
StyleKnee-length
No. of Pockets2
Collar TypeNotched Lapel
Model10029
Seam TypeSerged
Size (English) Chest39 to 41 in.
Certifications/ComplianceNFPA 99
For Use With (Application)Laboratories, Remediation, Decontamination, Pesticides
Length (English)38.5 in.
Length (Metric)97.79 cm
Construction1.6 oz. Fabric
Unit SizeCase