Reusable Small Animal Feeding Needles: Straight
Reusable Small Animal Feeding Needles: Straight
Actual product may vary
Reusable Small Animal Feeding Needles: Straight
Precision-assembled stainless-steel feeding needles ideal for feeding and dosing
Have Questions?
Catalog NumberDiameter (English) BallGaugeLength (English)
FIS01-290-3B0.088 in.20G1.5 in.
Catalog number FIS01-290-3B
Price (SGD)
2,401.00
Pack
Add to cart
Diameter (English) Ball:
0.088 in.
Gauge:
20G
Length (English):
1.5 in.
Price (SGD)
2,401.00
Pack
Add to cart
  • For the intubation of laboratory animals
  • Straight, curved tubing styles
  • Many standard sizes available, from small to large
  • Straight
  • Autoclavable
Small Animal Needles
  • For animals 0.7 oz. to 1 lb., 5 oz. (20 to 600g)
Specifications
AutoclavableAutoclavable
Product TypeStraight Animal Feeding Needle
MaterialStainless Steel
DisposableReusable
For Use With (Application)Small animal, Ideal for feeding and dosing
Gauge20G
Length (English)1.5 in.
Length (Metric)0.38 cm
Diameter (English) Ball0.088 in.
Diameter (Metric) Ball0.023 cm
Unit SizePack