Reusable Silica Gel Desiccant Bag
Actual product may vary
Fisherbrand
Reusable Silica Gel Desiccant Bag
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
Have Questions?
Catalog NumberCapacity (Metric)
FIS01-259-109350 g
Catalog number FIS01-259-109
Price (SGD)
85.00
Each
Capacity (Metric):
350 g
Price (SGD)
85.00
Each
  • Durable microfiber bag securely contains beads eliminating the risk of leakage and is unaffected by moisture
  • Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
  • Saturated gel can be regenerated by microwaving bag for 6 minutes
  • May be regenerated repeatedly without degradation
  • Arrives individually sealed in moisture-proof pouch
  • Available in multiple sizes to fit most desiccators
Specifications
Diameter (Metric) Outer5 cm
DescriptionDurable Microfiber Pouch
MaterialSilica Gel
Height (Metric)19 cm
Height (English)7.5 in.
Width (Metric)5 cm
Width (English)2 in.
Length (Metric)10 cm
Length (English)4 in.
Capacity (Metric)350 g
Capacity (English)0.75 lb.
Product TypeDesiccant Bag
Unit SizeEach