DuoSeal™ Pump Oil
Actual product may vary
Welch
DuoSeal™ Pump Oil
For use in DuoSeal High-Vacuum Pumps
Have Questions?
Catalog NumberVolume (Metric)
FIS01-104A1 L
Catalog number FIS01-104A
Price (SGD)
118.00
Each
Volume (Metric):
1 L
Price (SGD)
118.00
Each
For use in DuoSeal High-Vacuum Pumps
For use in DuoSeal High-Vacuum Pumps. One gallon.Specially fractionate oil has properties enabling highest performance for DuoSeal pumps
  • Low vapor Pressure
  • High Viscosity
Specifications
Product TypeVaccum Pump Oil
For Use With (Application)Provides the lubrication that helps enable renowned DuoSeal™ pump durability and ensure the highest vacuum performance
For Use With (Equipment)DuoSeal™ Belt-driven pumps
Quantity1 Ea.
Volume (English)0.26 gal.
Volume (Metric)1 L
Unit SizeEach