PYREX™ Reducing Glass Joint Adapters
Actual product may vary
Fisherbrand
PYREX™ Reducing Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
Have Questions?
Catalog NumberStandard Taper Joint No.Length (English)
FIS01-035D19/38 Inner Bottom, 24/40 Outer Top4.13 in.
Catalog number FIS01-035D
Price (SGD)
-
Standard Taper Joint No.:
19/38 Inner Bottom, 24/40 Outer Top
Length (English):
4.13 in.
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
With an outer standard taper joint at the top and a smaller inner standard taper joint at the bottom.
Specifications
For Use With (Equipment)For attaching apparatus with one joint size to equipment having a different size joint
Length (English)4.13 in.
Length (Metric)105 mm
MaterialBorosilicate Glass
Product TypeReducing Adapter
Standard Taper Joint No.19/38 Inner Bottom, 24/40 Outer Top
Unit SizePack