Welch Ilmvac Diaphragm Pump MPC 201T
Welch Ilmvac Diaphragm Pump MPC 201T
Actual product may vary
Welch Ilmvac Diaphragm Pump MPC 201T
Handle corrosive and aggressive vapor without damage. Fisher Scientific™ Diaphragm Pump MPC 201T is chemically resistantant and ideal for applications involving aggressive solvents and acidic vapors. The diaphragms and all gas- contaminated parts are made from PTFE and PTFE compound.
Have Questions?
Catalog number FILFB65454
Price (SGD)
-
  • Three stage chemically resistant diaphragm pumps
  • Quiet operation
  • >10,000 hour service intervals
  • Low capital and spare parts costs
  • Wetted parts from PTFE or PTFE compounds
  • Self cleaning valve design
  • Simplest maintenance
  • Compact modular design
  • Oil free reducing down time and operating costs
  • Substantially lower operating cost than water jet vacuum pumps
Applications:
  • For oil free applications within the rough vacuum range
  • Chemically resistant for use with solvent and acid vapours
  • Economical and ecologically sound alternative to water jet vacuum pumps
  • Vacuum filtration
  • Vacuum concentration
  • Drying ovens
  • Molecular distillation
Specifications
Motor Speed2/2.2 m3h
Certifications/ComplianceCE marked, UL/CSA approved
Electrical Requirements230 V 50/60 Hz
Depth (Metric)26 cm
MaterialPTFE, PTFE Compoud
Wattage90 w
Weight (English)22.7 lb.
Weight (Metric)10.3 kg
Flow Rate2 L/min.
Frequency50/60 Hz
Overall Height (English)5.9 in.
Overall Height (Metric)15 cm
Overall Length (Metric)260 mm
Overall Width (English)7.8 in.
Overall Width (Metric)20 cm
Maximum Pressure<2 mbar